EMAIL THIS PAGE TO A FRIEND
Tropical gastroenterology : official journal of the Digestive Diseases Foundation
Indispensable utility of intra-arterial calcium stimulation and hepatic venous sampling in a 7mm insulinoma with poor imaging localization.
PMID 22803305