EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical constituents from a portion of ethanolicextract of Saussurea lappa roots].
PMID 22803366
Abstract
To study chemical constituents from the roots of Saussurea lappa. Chemical constituents were separated and purified by various techniques such as silica gel column chromatography, Sephadex LH-20 and reversed phase RP-18 column chromatography. Their structures were identified on the basis of spectral data. Seventeen compounds were separated and identified as ascleposide E(1), (+)-1-hydroxypinoresinol-4"-O-methyl ester-4'-beta-D-glucopyranoside (2), (+)-1 -hydroxypinoresinol-4"-O-beta-D-glucopyranoside(3), (+)-1-hydroxypinoresinol-1-O-P-D-glucopyranoside (4), phenyl-beta-D-glucopyranoside (5), benzyl-beta-D-glucopyranoside (6), n-butyl-beta-D-glucopyranoside (7), ilicic alcohol (8), beta-cyclocostunolide (9), reynosin (10), 11beta, 13-dihydroreynosin (11), arbusculin A(12), 1beta-hydroxy-arbusculin A (13), santamarin (14), dehydrocostuslactone (15), 11beta, 13-dihydro-3-epizaluzanin C(16)and costunolide (17). Compounds 1-2 were separated from this genus for the first time,and compounds 3,5-7 were separated from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0417
Costunolide, ≥97% (HPLC)
C15H20O2