EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Nano-sized manganese oxide: a proposed catalyst for water oxidation in the reaction of some manganese complexes and cerium(IV) ammonium nitrate.
PMID 22806229
Abstract
According to UV-visible spectroscopy, X-ray diffraction spectrometry, dynamic light scattering, Fourier transform infrared spectroscopy, electron paramagnetic resonance spectroscopy, transmission electron microscopy, scanning electron microscopy and X-ray photoelectron spectroscopy, nano-sized manganese oxides are proposed as active catalysts for water oxidation in the reaction of some manganese complexes and cerium(IV) ammonium nitrate.