EMAIL THIS PAGE TO A FRIEND
Human psychopharmacology
Plasma fluvoxamine levels and OCD symptoms/response in adult patients.
PMID 22806821
Abstract
In this study, we explored the possible relationships between plasma fluvoxamine levels and clinical features and/or response in adult obsessive-compulsive disorder (OCD) patients treated with this drug for 6 months. Twenty OCD outpatients of both sexes who were already taking fluvoxamine (mean dose ± SD: 216.7 ± 86.2) for at least 4 weeks were included in the study. The severity of OCD was assessed by means of the Yale-Brown obsessive-compulsive scale (Y-BOCS). The fluvoxamine plasma levels were measured by high-performance liquid chromatography analysis. All evaluations were performed after 4 weeks (t1) and 6 months (t2) of fluvoxamine intake. The plasma levels of fluvoxamine remained stable at the two assessment times, with no sex-related differences. Sixteen (80%) patients responded to treatment as shown by the significant (>35%) decrease of the Y-BOCS total score. Men's compulsions improved more than those of women. Significant and positive correlations were detected between fluvoxamine plasma levels at t1 and t2 and the difference (delta) of the Y-BOCS total and compulsion subscale scores between t1 and t2. Another significant, albeit negative, correlation was measured between the difference of the compulsion subscale score and the difference of fluvoxamine levels at t1 and t2. These findings underline the potential importance of evaluating fluvoxamine plasma levels in OCD and their relationships with the clinical response that may be gender-related on specific symptoms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000853
Fluvoxamine for system suitability, European Pharmacopoeia (EP) Reference Standard
C15H21F3N2O2 · C4H4O4
F2802
Fluvoxamine maleate, solid
C15H21F3N2O2 · C4H4O4