EMAIL THIS PAGE TO A FRIEND
International journal of STD & AIDS
Successful treatment of giant condyloma acuminatum with combination retinoid and interferon-γ therapy.
PMID 22807543
Abstract
Giant condyloma acuminatum, or Buschke-Löwenstein tumour, is a unique variant of anogenital warts. It is characterized by locally aggressive behaviour but rarely metastasizes. Many treatment strategies such as radical surgery, radiation and chemotherapy have been used to treat it but their efficacy is often poor and the recurrence rates are high. We report a case of 16-year-old girl who was treated with oral retinoid combined with intramuscular interferon-γ. All lesions cleared within three months. During a follow-up period of more than two years, no recurrence has developed. This relatively painless, non-scarring treatment may represent a novel therapeutic option.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0225000
Acitretin, European Pharmacopoeia (EP) Reference Standard
C21H26O3