EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Markers of transmembrane and energy exchange in cells of terminal placental villi in spontaneous and induced pregnancy.
PMID 22808512
Abstract
We performed a comparative morphological study of the placentas in spontaneous and induced pregnancy. Immunohistochemical analysis revealed pronounced decrease in the expression of markers of transmembrane (α-SNAP 23, annexin 3) and energy (ferritin light chain, ATP5J) in placental terminal villi.
Related Materials