EMAIL THIS PAGE TO A FRIEND
Expert opinion on drug metabolism & toxicology
ADMET considerations for restless leg syndrome drug treatments.
PMID 22808933
Abstract
Restless legs syndrome (RLS) is a common neurological disorder that might impair nocturnal rest causing decreased alertness, depressed mood, reduced job performance, and poor quality of life. In patients affected by severe RLS, a pharmacological treatment is mandatory. The present review is based on a search using PubMed from 1994 to 2012. It is focused on the Absorption, Distribution, Metabolism, Elimination and Toxicology (ADMET) characteristics of the most used medications for RLS. In particular, the ADMET characteristics of dopaminergic agents, anticonvulsants able to improve neuropathic pain, and iron were discussed. Clinical trials have showed that non-ergolic dopamine agonists are efficacious and safe for patients affected by moderate to severe idiopathic RLS. However, no head-to-head study has compared the long-term effects of the three dopamine agonists approved by the FDA for RLS (ropinirole, pramipexole, and rotigotine). Moreover, further studies should investigate the extended-release formulation of ropinirole and pramipexole in RLS patients affected by all day long distressing symptoms. A standardized treatment for symptomatic forms of RLS is lacking. Randomized, placebo-controlled trials should be performed at least in RLS patients with peripheral neuropathic and chronic kidney disease. Concerning RLS due to iron deficiency, a head-to-head study comparing efficacy, safety and compliance of oral iron versus intravenous one seems to be needed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2530
Ropinirole hydrochloride, powder, ≥98% (HPLC)
C16H24N2O · HCl
R9281
Rotigotine hydrochloride, ≥98% (HPLC)
C19H25NOS·HCl