EMAIL THIS PAGE TO A FRIEND
Vestnik otorinolaringologii
[The experience with the supervision and treatment of the patients presenting with external otitis].
PMID 22810639
Abstract
This clinical study was designed to estimate the efficacy and tolerability of the locally applied Pimafucort preparation for the treatment of diffuse otitis of bacterial and mycotic origin. The results of dynamic bacteriological investigation in conjunction with clinical observations give reason to recommend the local therapy as the sole treatment of uncomplicated forms of diffuse external otitis.