EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Pyridofuran substituted pyrimidine derivatives as HCV replication (replicase) inhibitors.
PMID 22814211
Abstract
Introduction of nitrogen atom into the benzene ring of a previously identified HCV replication (replicase) benzofuran inhibitor 2, resulted in the discovery of the more potent pyridofuran analogue 5. Subsequent introduction of small alkyl and alkoxy ligands into the pyridine ring resulted in further improvements in replicon potency. Replacement of the 4-chloro moiety on the pyrimidine core with a methyl group, and concomitant monoalkylation of the C-2 amino moiety resulted in the identification of several inhibitors with desirable characteristics. Inhibitor 41, from the monosubstituted pyridofuran and inhibitor 50 from the disubstituted series displayed excellent potency, selectivity (GAPDH/MTS CC(50)) and PK parameters in all species studied, while the selectivity in the thymidine incorporation assay (DNA·CC(50)) was low.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B8002
2,3-Benzofuran, 99%
C8H6O