EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
PERTINENT (a substudy of the EUROPA trial): a persistent legacy.
PMID 22815531
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0094
Perindopril erbumine
C19H32N2O5 · C4H11N