EMAIL THIS PAGE TO A FRIEND
Natural product communications
Cis-aconitic anhydride ethyl ester and phenolic compounds from the seeds of Alisma orientale.
PMID 22816308
Abstract
From the seeds of Alisma orientale, cis-aconitic anhydride ethyl ester and cis-2,4,5-trihydroxycinnamic acid were isolated, together with nine known phenolic compounds and a megastigmane sesquiterpene. All compounds are reported for the first time from Alisma species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
217808
cis-Aconitic anhydride, 95%
C6H4O5