EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Novel layer-by-layer procedure for making nylon-6 nanofiber reinforced high strength, tough, and transparent thermoplastic polyurethane composites.
PMID 22817392
Abstract
We highlight a novel composite fabrication method based on solution casting, electrospinning, and film stacking for preparing highly transparent nylon-6 nanofiber reinforced thermoplastic polyurethane (TPU) composite films. The procedure is simple and can be extended to the other thermoplastics. The morphology of fiber/matrix interface and the properties of composite films were also investigated. The method led to a significant reinforcement in mechanical properties of TPU like tensile strength, E modulus, strain, and toughness just using very small amounts of nylon fibers (about 0.4-1.7 wt %; 150-300 nm diameter). The enhanced mechanical properties were achieved without sacrificing optical properties like transparency of TPU.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2204
ε-Caprolactam, 99%
C6H11NO
181110
Nylon 6, pellets