EMAIL THIS PAGE TO A FRIEND
Cancer radiotherapie : journal de la Societe francaise de radiotherapie oncologique
[Evaluation and management of acute radiation dermatitis].
PMID 22818410
Abstract
Acute radiation dermatitis remains one of the most commonly observed side effect during radiation therapy leading to complication such as superinfection or treatment disruption. Its management is characterized by a great heterogeneity. Few strategies have demonstrated a benefit in preventing radiation dermatitis, which relies mostly on decreasing dose delivered to the skin and skin care practices. Simple emollients and use of topical steroids can be useful in early stages. The singularity of the skin toxicity seen with cetuximab and radiotherapy warrants a specific grading system and distinctive clinical treatment with use of antibiotics.
Related Materials