EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
(+)-Pinoresinol is a putative hypoglycemic agent in defatted sesame (Sesamum indicum) seeds though inhibiting α-glucosidase.
PMID 22818971
Abstract
Defatted sesame seeds have been reported for hypoglycemic effect in mice and T2DM women. An attempted to identify active components responsible for this effect was conducted using α-glucosidase-guided fractionation, resulting in the isolation of various lignans. Of compounds isolated, only (+)-pinoresinol showed inhibitory activity against rat intestinal maltase with an IC(50) value of 34.3 μM. The kinetic study indicated that enzymatic hydrolysis of maltose is inhibited by (+)-pinoresinol through competitive and noncompetitive manners. However, a lower dissociation constant (k(i) 288 M) of EI complex suggested that competitive inhibition is predominant over noncompetitive mode (k'(i) 1342 M).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06709
Pinoresinol, analytical standard
C20H22O6
40674
Pinoresinol, ≥95.0% (HPLC)
C20H22O6