EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Understanding trans platinum complexes as potential antitumor drugs beyond targeting DNA.
PMID 22819649
Abstract
This focused review is devoted to our advances in developing trans platinum complexes as antitumoral complexes, in particular those bearing aliphatic amines. It includes some fundamental aspects of the understanding of non conventional metallodrug mechanism. The perspective is arranged by those fundamental aspects and within each section only the most successful examples have been included and also those in which clearly SAR's (structure activity relationships) have been observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
373214
Platinum, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Pt
373230
Platinum, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Pt
349372
Platinum, foil, thickness 1.0 mm, 99.99% trace metals basis
Pt
267260
Platinum, foil, thickness 0.5 mm, 99.99% trace metals basis
Pt
305189
Platinum, foil, thickness 0.5 mm, 99.9% trace metals basis
Pt
349321
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
Pt
327425
Platinum, foil, thickness 0.25 mm, 99.9% trace metals basis
Pt
349348
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
Pt
327433
Platinum, foil, thickness 0.125-0.135 mm, 99.9% trace metals basis
Pt
267252
Platinum, foil, thickness 0.1 mm, 99.99% trace metals basis
Pt
349380
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
Pt
349356
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
Pt
303798
Platinum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Pt
349364
Platinum, foil, thickness 0.025 mm, 99.99% trace metals basis
Pt
267244
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
Pt
298107
Platinum, gauze, 52 mesh, 99.9% trace metals basis
Pt
298093
Platinum, gauze, 100 mesh, 99.9% trace metals basis
Pt
685453
Platinum, nanopowder, <50 nm particle size (TEM)
Pt
204048
Platinum, powder (coarse), 99.99% trace metals basis
Pt
204013
Platinum, powder, 99.995% trace metals basis
Pt
327476
Platinum, powder, ≤10 μm, 99.9% trace metals basis
Pt
327484
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
Pt
267155
Platinum, sponge, ≥99.9% trace metals basis
Pt
444685
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
Pt
349399
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
Pt
267236
Platinum, wire, diam. 1.0 mm, 99.99% trace metals basis
Pt
327492
Platinum, wire, diam. 1.0 mm, 99.9% trace metals basis
Pt
267201
Platinum, wire, diam. 0.5 mm, 99.99% trace metals basis
Pt
267228
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis
Pt
267171
Platinum, wire, diam. 0.25 mm, 99.99% trace metals basis
Pt