EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Charge-order driven proton arrangement in a hydrogen-bonded charge-transfer complex based on a pyridyl-substituted TTF derivative.
PMID 22820668
Abstract
A unique N(+)-H···N hydrogen-bonded (H-bonded) dimer motif based on partially oxidized pyridyl-substituted TTF was constructed in the charge-transfer complex. The charge ordering in the TTF column by the charge disproportionation in the dimer regulates the arrangement of the H-bonded proton, evidencing the proton-electron coupled state.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183180
Tetrathiafulvalene, 97%
C6H4S4