EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Solvent-stabilized y-type oxotitanium phthalocyanine photoconductive nanoparticles: preparation and application in single-layered photoreceptors.
PMID 22821758