EMAIL THIS PAGE TO A FRIEND
Saudi journal of gastroenterology : official journal of the Saudi Gastroenterology Association
Increased protein carbonylation and decreased antioxidant status in anemic H. pylori infected patients: effect of treatment.
PMID 22824768
Abstract
Collective evidences suggest the causal association of Helicobacter pylori infection with iron deficiency anemia. Generation of free radicals against this bacterium can lead to turbulence in oxidative-antioxidative system. This study was undertaken to evaluate the marker of oxidative protein injury, protein carbonylation, and total antioxidant status in anemic H. pylori-infected patients and to observe the alteration in them after treatment for 1 month with oral ferrous sulfate and anti-H. pylori therapy. Twenty anemic H. pylori-infected patients were randomly divided into 2 groups. The H. pylori-infected patients in Group I received both iron supplementation and anti-H pylori therapy, whereas patients in Group II received only the iron supplementation. Fifteen healthy volunteers served as controls. All the study parameters were estimated after 1 month of the treatment. Protein carbonylation and total antioxidant status were estimated using colorimetric method. Hematologic parameters were evaluated using Sysmex-K-100 automated cell counter. In anemic H. pylori-infected patients, the protein carbonyls (PCOs) were significantly increased, whereas the total antioxidant status, iron, hemoglobin, and ferritin levels were significantly decreased compared with the controls. In Group I, while the PCOs level decreased significantly, there was a significant increase in the total antioxidant status, iron, hemoglobin, and ferritin levels after 1 month. No significant alterations were noted in the levels of PCOs, total antioxidant status, iron, hemoglobin, or ferritin in Group II patients after 1 month of the treatment. The findings from this study indicate that treatment for both anemia and H. pylori infections is required for lowering the oxidative stress markers, which synergistically bring about an appropriate correction of anemia soon in these patients.