EMAIL THIS PAGE TO A FRIEND
Cardiovascular drugs and therapy
HDL- cholesterol: a nut too hard to crack?
PMID 22825352
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0170
Torcetrapib, ≥98% (HPLC)
C26H25F9N2O4