EMAIL THIS PAGE TO A FRIEND
Journal of environmental monitoring : JEM
Characterizing the intrinsic bioremediation potential of 1,4-dioxane and trichloroethene using innovative environmental diagnostic tools.
PMID 22825917
Abstract
An intrinsic biodegradation study involving the design and implementation of innovative environmental diagnostic tools was conducted to evaluate whether monitored natural attenuation (MNA) could be considered as part of the remedial strategy to treat an aerobic aquifer contaminated with 1,4-dioxane and trichloroethene (TCE). In this study, advanced molecular biological and stable isotopic tools were applied to confirm in situ intrinsic biodegradation of 1,4-dioxane and TCE. Analyses of Bio-Trap® samplers and groundwater samples collected from monitoring wells verified the abundance of bacteria and enzymes capable of aerobically degrading TCE and 1,4-dioxane. Furthermore, phospholipid fatty acid analysis with stable isotope probes (PLFA-SIP) of the microbial community validated the ability for microbial degradation of TCE and 1,4-dioxane. Compound specific isotope analysis (CSIA) of groundwater samples for TCE resulted in δ(13)C values that indicated likely biodegradation of TCE in three of the four monitoring wells sampled. Results of the MNA evaluation showed that enzymes capable of aerobically degrading TCE and 1,4-dioxane were present, abundant, and active in the aquifer. Taken together, these results provide direct evidence of the occurrence of TCE and 1,4-dioxane biodegradation at the study site, supporting the selection of MNA as part of the final remedy at some point in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
360481
1,4-Dioxane, ACS reagent, ≥99.0%
C4H8O2
34857
1,4-Dioxane, for HPLC, ≥99.5%
C4H8O2
D201863
1,4-Dioxane, ReagentPlus®, ≥99%
C4H8O2
33147
1,4-Dioxane, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C4H8O2
676934
1,4-Dioxane, ACS reagent, ≥99.0%
C4H8O2
CRM48367
1,4-Dioxane solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL
C4H8O2
551368
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D)
C4H8O2
736171
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 5 mm × 8 in.
C4H8O2
611905
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 5 mm × 8 in.
C4H8O2
704806
1,4-Dioxane solution, NMR reference standard, 1 mM in D2O ("100", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
704814
1,4-Dioxane solution, NMR reference standard, 10 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
704822
1,4-Dioxane solution, NMR reference standard, 0.1 mM in D2O ("100%", 99.96 atom % D), NMR tube size 5 mm × 7 in.
C4H8O2
703354
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 6.5 mm × 8 in.
C4H8O2
684228
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 10 mm × 8 in.
C4H8O2
631868
1,4-Dioxane solution, NMR reference standard, 5 mM in chloroform-d (99.8 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
631833
1,4-Dioxane solution, NMR reference standard, 5 mM in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2
612073
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), chromium(III) acetylacetonate 5 mg/mL, NMR tube size 3 mm × 8 in.
C4H8O2
612111
1,4-Dioxane solution, NMR reference standard, 40% in benzene-d6 (99.6 atom % D), NMR tube size 3 mm × 8 in.
C4H8O2