EMAIL THIS PAGE TO A FRIEND
Zhurnal evoliutsionnoi biokhimii i fiziologii
[Reversible lupininin inhibitors of cholinesterases of mammalian blood and of optical ganglia of individuals of the commander squid Berryteuthis magister from different zones of species areal].
PMID 22827020
Abstract
Arylsulfoesters and carbonic lupinin esters are studied for the first time as reversible inhibitors of mammalian blood cholinesterases. Studied in detail is sensitivity of cholinesterases to mono- and bislupinin inhibitors in Commander squid individuals from different habitation zones.