EMAIL THIS PAGE TO A FRIEND
Dermatitis : contact, atopic, occupational, drug
Allergic contact dermatitis syndrome from bufexamac for nursing infant.
PMID 22828265
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0760
Bufexamac, analytical standard
C12H17NO3