EMAIL THIS PAGE TO A FRIEND
Water science and technology : a journal of the International Association on Water Pollution Research
Treatment of ametryn in wastewater by a hybrid MBR system: a lab-scale study.
PMID 22828312
Abstract
Agricultural discharge of herbicides to the Great Barrier Reef (GBR) poses significant threat to the marine ecosystem. This study evaluates the performance of a hybrid treatment system consists of a membrane bioreactor (MBR), UV disinfection unit and a granular activated carbon (GAC) column in treating ametryn which is one of the major herbicides in agricultural discharges. While the MBR alone removes only 40% of ametryn at a hydraulic retention time of 7.8 h, the hybrid system removed ametryn to below detection levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N11064
Ametryn, analytical standard
C9H17N5S
45321
Ametryn, PESTANAL®, analytical standard
C9H17N5S