EMAIL THIS PAGE TO A FRIEND
Clinical research in cardiology : official journal of the German Cardiac Society
Pro-thrombotic condition in a woman with peripartum cardiomyopathy treated with bromocriptine and an Impella LP 2.5 heart pump.
PMID 22829017