EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Identification of a novel pathway of transforming growth factor-β1 regulation by extracellular NAD+ in mouse macrophages: in vitro and in silico studies.
PMID 22829588
Abstract
Extracellular β-nicotinamide adenine dinucleotide (NAD(+)) is anti-inflammatory. We hypothesized that NAD(+) would modulate the anti-inflammatory cytokine Transforming Growth Factor (TGF)-β1. Indeed, NAD(+) led to increases in both active and latent cell-associated TGF-β1 in RAW 264.7 mouse macrophages as well as in primary peritoneal macrophages isolated from both C3H/HeJ (TLR4-mutant) and C3H/HeOuJ (wild-type controls for C3H/HeJ) mice. NAD(+) acts partially via cyclic ADP-ribose (cADPR) and subsequent release of Ca(2+). Treatment of macrophages with the cADPR analog 3-deaza-cADPR or Ca(2+) ionophores recapitulated the effects of NAD(+) on TGF-β1, whereas the cADPR antagonist 8-Br-cADPR, Ca(2+) chelation, and antagonism of L-type Ca(2+) channels suppressed these effects. The time and dose effects of NAD(+) on TGF-β1 were complex and could be modeled both statistically and mathematically. Model-predicted levels of TGF-β1 protein and mRNA were largely confirmed experimentally but also suggested the presence of other mechanisms of regulation of TGF-β1 by NAD(+). Thus, in vitro and in silico evidence points to NAD(+) as a novel modulator of TGF-β1.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7344
Cyclic adenosine diphosphate-ribose, ≥90% (HPLC), lyophilized powder
C15H21N5O13P2