EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
A robust metal-organic framework constructed from alkoxo-bridged binuclear nodes and hexamethylenetetramine spacers: crystal structure and sorption studies.
PMID 22830491
Abstract
A neutral 3D metal-organic framework, (3)(∞)[Cu(2)(mand)(2)(hmt)]·H(2)O (1), was constructed from binuclear Cu(2)O(2) alkoxo-bridged nodes, generated by the doubly deprotonated mandelic acid. The nodes are connected by hexamethylenetetramine (hmt) spacers, which act as biconnective bridging ligands, and by carboxylato groups. Channels are observed along the crystallographic c axis. The water molecules from the channels can be easily removed, preserving the architecture of the crystal, which is stable up to 280 °C. The Langmuir surface area was found to be 610 m(2) g(-1). The sorption ability of 1 was investigated using H(2) and CO(2).