EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Extraction-photometric analysis of low levels of hem-ditret-butylperoxy-3,3,5-trimethylcyclohexane peroxide in in water solutions].
PMID 2283083
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
479896
Luperox® 231, 1,1-Bis(tert-butylperoxy)-3,3,5-trimethylcyclohexane, 92%
C17H34O4