EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Effect of enzymatic deamidation of soy protein by protein-glutaminase on the flavor-binding properties of the protein under aqueous conditions.
PMID 22831747
Abstract
The effect of the enzymatic deamidation by protein-glutaminase (PG) on flavor-binding properties of soy protein isolate (SPI) under aqueous conditions was evaluated by a modified equilibrium dialysis (ultrafiltration) technique. Binding parameters, such as number of binding sites (n) and binding constants (K), were derived from Klotz plots. The partial deamidation of SPI by PG (43.7% degree of deamidation) decreased overall flavor-binding affinity (nK) at 25 °C for both vanillin and maltol by approximately 9- and 4-fold, respectively. The thermodynamic parameters of binding indicated that the flavor-protein interactions were spontaneous (negative ΔG°) and that the driving force of the interactions shifted from entropy to enthalpy driven as a result of deamidation. Deamidation of soy protein caused a change in the mechanism of binding from hydrophobic interactions or covalent bonding (Schiff base formation) to weaker van der Waals forces or hydrogen bonding.