EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Zeolite Y encapsulated with Fe-TiO2 for ultrasound-assisted degradation of amaranth dye in water.
PMID 22831996
Abstract
A new heterogeneous catalyst for sonocatalytic degradation of amaranth dye in water was synthesized by introducing titania into the pores of zeolite (NaY) through ion exchange method while Fe (III) was immobilized on the encapsulated titanium via impregnation method. XRD results could not detect any peaks for titanium oxide or Fe(2)O(3) due to its low loading. The UV-vis analysis proved a blue shift toward shorter wavelength after the loading of Ti into NaY while a red shift was detected after the loading of Fe into the encapsulated titanium. Different reaction variables such as TiO(2) content, amount of Fe, pH values, amount of hydrogen peroxide, catalyst loading and the initial dye concentration were studied to estimate their effect on the decolorization efficiency of amaranth. The maximum decolorization efficiency achieved was 97.5% at a solution pH of 2.5, catalyst dosage of 2 g/L, 20 mmol/100 mL of H(2)O(2) and initial dye concentration of 10 mg/L. The new heterogeneous catalyst Fe/Ti-NaY was a promising catalyst for this reaction and showed minimum Fe leaching at the end of the reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1016
Amaranth, Dye content 85-95 %
C20H11N2Na3O10S3
87612
Amaranth, analytical standard
C20H11N2Na3O10S3