EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Enantio- and diastereocontrolled conversion of chiral epoxides to trans-cyclopropane carboxylates: application to the synthesis of cascarillic acid, grenadamide and L-(-)-CCG-II.
PMID 22832742
Abstract
An efficient high yielding improved method for the enantio- and diastereoselective cyclopropanation of chiral epoxides using triethylphosphonoacetate and base (Wadsworth-Emmons cyclopropanation) is reported. The utility of this protocol is illustrated by concise and practical synthesis of cascarillic acid, grenadamide and L-(-)-CCG-II, a cyclopropane containing natural products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C195
(2S,3R,4S)-α-(Carboxycyclopropyl)glycine, solid
C6H9NO4