EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Nanoscale manganese oxide within Faujasite zeolite as an efficient and biomimetic water oxidizing catalyst.
PMID 22833185
Abstract
Nanoscale manganese oxides within Faujasite zeolite have been synthesized with a simple method and characterized by scanning electron microscopy, X-ray diffraction spectrometry, N(2) adsorption-desorption isotherms, transmission electron microscopy, and atomic absorption spectroscopy. These oxides showed efficient water oxidizing activity in the presence of cerium(IV) ammonium nitrate as a non-oxo transfer oxidant.