EMAIL THIS PAGE TO A FRIEND
Molecular biology of the cell
Interplay between MEK-ERK signaling, cyclin D1, and cyclin-dependent kinase 5 regulates cell cycle reentry and apoptosis of neurons.
PMID 22833568
Abstract
In response to neurotoxic signals, postmitotic neurons make attempts to reenter the cell cycle, which results in their death. Although several cell cycle proteins have been implicated in cell cycle-related neuronal apoptosis (CRNA), the molecular mechanisms that underlie this important event are poorly understood. Here, we demonstrate that neurotoxic agents such as β-amyloid peptide cause aberrant activation of mitogen-activated kinase kinase (MEK)-extracellular signal-regulated kinase (ERK) signaling, which promotes the entry of neurons into the cell cycle, resulting in their apoptosis. The MEK-ERK pathway regulates CRNA by elevating the levels of cyclin D1. The increase in cyclin D1 attenuates the activation of cyclin-dependent kinase 5 (cdk5) by its neuronal activator p35. The inhibition of p35-cdk5 activity results in enhanced MEK-ERK signaling, leading to CRNA. These studies highlight how neurotoxic signals reprogram and alter the neuronal signaling machinery to promote their entry into the cell cycle, which eventually leads to neuronal cell death.