EMAIL THIS PAGE TO A FRIEND
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova
[Apoptosis of the hypothalamus neurosecretory cells in stress and ageing: the role of immune modulators].
PMID 22834344
Abstract
There is assumption about active role of immune modulators in cell death process. The involvement of interferon-alpha and cycloferon in apoptosis regulation of hypothalamic neurons of mice during stress and aging was studied. We determined the expression of apoptosis markers (Bcl-2, Mcl-1, Bax) in comparison with apoptosis level. We have found that immune modulators suppress activity of nonapeptidergic neurons. Thus, interferon-alpha treatment reduces synthesis of Bcl-2; cycloferon treatment inhibits expression of Bax and Bcl-2. So the role of immune modulators in neuron apoptosis depends on the stage of ontogenesis and type of immune modulator. Cycloferon is able to reduce the level of age-dependent apoptosis of neurons in aging, but under stress condition both interferon-alpha and cycloferon act as protectors of cell death.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17927
9-Oxo-10(9H)-acridineacetic acid, for HPLC derivatization, ≥99.0% (T)
C15H11NO3