EMAIL THIS PAGE TO A FRIEND
Archiv fur Kriminologie
[Analysis with the Combur-Test--special aspects in forensic trace examination].
PMID 22834362
Abstract
The Combur Test is a ready-made and easy-to-use pretest for blood. It is based on the oxidation of tetramethylbenzidine (TMB), which is catalysed by haemoglobin and its derivatives. Despite its high sensitivity, there are many known substances which are responsible for false positive and false negative test results. On the basis of experiments of our own, case reports and the pertinent literature special aspects of the application of the Combur Test in the forensic routine case work are discussed.