EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Unexpected one-pot synthesis of highly conjugated pentacyclic diquinoid compounds.
PMID 22834705
Abstract
A new class of pentacyclic diquinoid compounds has been synthesized with a facile one-pot reaction of two molecules of 2-hydroxynaphthoquinone and 1-bromoalkanes in the presence of ferrocene. These molecules were isolated as enol tautomers that exhibit intramolecular hydrogen bond and extended electronic conjugation as proved by the intense absorption spectrum with a broad band between 400 and 600 nm. The spectroscopic and electrochemical characterization of this new class of compounds has been performed. One of the synthesized diquinoid derivatives showed a significant cytotoxicity with IC(50) values of 25-50 μM against Cisplatin-Resistant SKOV3 and colon carcinoma SW480 cell lines. The results of our study provide a valuable tool to a one-pot synthesis of highly conjugated polyquinones, analogous to important biological systems, with significant antitumoral activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H46805
2-Hydroxy-1,4-naphthoquinone, 97%
C10H6O3