EMAIL THIS PAGE TO A FRIEND
Journal of aquatic animal health
Oxytetracycline pharmacokinetics in rainbow trout during and after an orally administered medicated feed regimen.
PMID 22838083
Abstract
The pharmacokinetic-pharmacodynamic predictor of antimicrobial activity for tetracyclines is reported to be the area under the concentration-time curve at steady state (AUC(ss)) divided by the minimal inhibitory concentration of the targeted pathogen. Here, we estimate AUC(ss) values for oxytetracycline (OTC) in serum of rainbow trout Oncorhynchus mykiss by using a destructive sampling study design. Seventy-two rainbow trout were fed OTC-medicated feed at 74.7 +/- 1.5 mg/kg (mean +/- SD) body weight (BW) by oral gavage for 10 consecutive days. Serum was collected from nine fish at 1, 3, 6, 8, 10, 12, 15, and 22 d after dosing began. Serum OTC concentrations were measured by high-performance liquid chromatography with a 0.01-microg/mL limit of detection. The average OTC AUC(ss) was 29.2 microg x h/mL and was estimated using nonlinear mixed-effects modeling and bootstrap resampling techniques. The elimination half-life was estimated as 85.0 h, and the fraction of steady state achieved was estimated as 0.85. The calculated AUC(ss) (24.8 microg x h/mL) following 10 d of oral dosing with 75 mg OTC/kg BW was less than the estimated AUC(ss). Results suggest that the pharmacokinetics of OTC exposure, including the AUC(ss), is better evaluated by using multiday dosimetry than by using a standard single-dose protocol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O4636
Oxytetracycline dihydrate, Assay (anhydrous basis) 95.0%-102.0%
C22H24N2O9 · 2H2O