EMAIL THIS PAGE TO A FRIEND
Kardiologiia
[Remodeling of vascular bed in patients with arterial hypertension: possibilities of diagnostics and correction].
PMID 22839673
Abstract
Arterial hypertension determines the remodeling of the vascular bed at all levels, resulting in a high risk of developing cardiovascular complications. Recent studies performed on the basis of evidence-based medicine, demonstrated the high efficiency of the combination drug therapy with perindopril and amlodipine in preventing such complications. This may be due to the positive influence of a fixed combination of these drugs on the parameters of the state of the microcirculatory level of the vascular bed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0094
Perindopril erbumine
C19H32N2O5 · C4H11N