EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Modification of hydroxypropyl guar gum with ethanolamine.
PMID 22840030
Abstract
A new guar gum derivative containing amino group was synthesized through nucleophilic substitution of p-toluenesulfonate activated hydroxypropyl guar gum with ethanolamine. For the preparation of p-toluenesulfonate esters hydroxypropyl guar gum, the results showed that the reaction rate was optimal at 25 °C and the reaction could reach equilibrium state when it was carried out for 10h at 25 °C. For the nucleophilic substitution of tosyl group with ethanolamine, the reaction was completed after 10h reaction at 50 °C. The structures of products were characterized by NMR and FT-IR spectroscopy. The results showed that the p-toluenesulfonate esters can be effectively substituted by ethanolamine to form the hydroxyethyl amino hydroxypropyl guar gum (EAHPG). The content of nitrogen of EAHPG was determined by acid-base titration and element analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4129 Guar