EMAIL THIS PAGE TO A FRIEND
Biomaterials
Multifunctional ZnPc-loaded mesoporous silica nanoparticles for enhancement of photodynamic therapy efficacy by endolysosomal escape.
PMID 22840227
Abstract
The cellular uptake and localization of photosensitizer-loaded nanoparticles have significant impact on photodynamic therapy (PDT) efficacy due to short lifetime and limited action radius of singlet oxygen. Herein, we develop poly(ethylene glycol) (PEG)- and polyethylenimine (PEI)-functionalized zinc(II) phthalocyanine (ZnPc)-loaded mesoporous silica nanoparticles (MSNs), which are able to distribute in the cytosol by endolysosomal escape. In this photosensitizer-carrier system (PEG-PEI-MSNs/ZnPc), ZnPc is a PDT agent; MSNs are the nanocarrier for encapsulating ZnPc; PEI facilitates endosomal escape; and PEG enhances biocompatibility. The as-synthesized PEG-PEI-MSNs/ZnPc have a high escape efficiency from the lysosome to the cytosol due to the "proton sponge" effect of PEI. Compared with the ZnPc-loaded MSNs, the phototoxicity of the PEG-PEI-MSNs/ZnPc is greatly enhanced in vitro. By measuring the mitochondrial membrane potential, a significant loss of >80% Δψm after treatment with PEG-PEI-MSNs/ZnPc-PDT is observed. It is further demonstrated that the ultra-efficient passive tumor targeting and excellent PDT efficacy are achieved in tumor-bearing mice upon intravenous injection of PEG-PEI-MSNs/ZnPc and the followed light exposure. We present here a strategy for enhancement of PDT efficacy by endolysosomal escape and highlight the promise of using multifunctional MSNs for cancer therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
341169
Zinc phthalocyanine, Dye content 97 %
C32H16N8Zn