EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and environmental safety
Sorption behavior of sulfamethazine on unamended and manure-amended soils and short-term impact on soil microbial community.
PMID 22840437
Abstract
In this study we investigated the sorption of sulfamethazine (SMZ) in two soils with different physico-chemical properties and the sulfonamide short-term influence on the microbial community structure and function. The presence of manure increased the SMZ sorption, the uppermost level being measured on soil with the lower pH and the higher manure content allowed by the Italian regulation. The sulfonamide desorption was hysteretic on both soils. SMZ addition to soil at the concentration of 53.6 μg/g had a significant short-term negative impact on readily culturable bacteria, potential metabolic activity (Biolog CLPP) and soil enzyme activity. Moreover, a shift of the culturable microbial populations towards a lower bacteria/fungi ratio was observed after SMZ addition. Despite the observed SMZ effects disappeared almost completely after 7 day, structural changes of microbial communities were still present in SMZ-treated soils. The results presented are remarkable since previous studies addressing the SMZ impact on soil microbial parameters failed to highlight any significant effect of the sulfonamide on microbial abundance and diversity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46802
Sulfamethazine, VETRANAL, analytical standard
C12H14N4O2S