EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Effects of long term exposure to the mycotoxin fumonisin B1 in p53 heterozygous and p53 homozygous transgenic mice.
PMID 22841953
Abstract
The fungal toxin fumonisin B1 (FB1) is a potential human carcinogen based on evidence of renal carcinogenicity in rats and hepatocarcinogenicity in mice. The toxicity and carcinogenicity of FB1 is linked to ceramide synthase inhibition. Based on this mechanism of action and on lack of evidence of genotoxicity, FB1 is considered a non-genotoxic carcinogen. The p53 heterozygous (p53+/-) mouse is a cancer-prone model used for carcinogenesis. The effects of chronic dietary FB1 exposure were characterized in p53+/- mice to confirm non-genotoxicity using a model which is more sensitive to genotoxic than non-genotoxic carcinogens and to clarify the relationship between p53 expression, altered sphingolipid metabolism, and FB1-induced carcinogenesis. Responses to FB1 were similar in p53+/- and p53+/+ mice after 26 weeks exposure to 0, 5, 50 or 150 mg FB1/kg diet, supporting a non-genotoxic mechanism of action. Hepatic adenomas and cholangiomas were observed in mice exposed to 150 mg/kg FB1. For a 10% increase in hepatic megalocytosis, the estimated 95% lower confidence limit of the benchmark dose (BMDL10) ranged from 0.15 and 1.11 mg FB1/kg bw/day. Based on similar responses in p53+/- and p53+/+ mice, p53 and related pathways play a secondary role in responses to FB1 toxicity and carcinogenesis.
Related Materials