EMAIL THIS PAGE TO A FRIEND
Polish journal of veterinary sciences
Activity of selected antioxidative enzymes in rats exposed to dimethoate and pyrantel tartrate.
PMID 22844700
Abstract
This study presents the results of research concerning the effect of single and combined application of pyrantel tartrate and dimethoate on selected antioxidative enzymes: catalase (CAT), superoxide dismutase (SOD) and glutathione peroxidase (GPx), in rat erythrocytes. Pyrantel tartrate was applied twice, at a dose of 85 mg/kg bw at a two week interval, i.e. on day 14 and 28 of the experiment, orally, in a water solution with a stomach tube. Dimethoate was administered with drinking water for 28 days at a dose of 25 mg/kg bw/day. It was found that pyrantel tartrate caused only small changes in the activity of the antioxidative enzymes under analysis. Subchronic exposure of rats to dimethoate caused a significant increase in the activity of CAT, SOD and GPx in erythrocytes, indicating the existence of strong oxidative stress. In combined intoxication, no significant effects of administering pyrantel tartrate on the activity of CAT, SOD and GPx was found in animals poisoned with dimethoate. The profile of changes was similar to that observed in rats exposed only to the organophosphorus insecticide. This may indicate a lack of interaction between the compounds used in the experiment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45449
Dimethoate, PESTANAL®, analytical standard
C5H12NO3PS2