EMAIL THIS PAGE TO A FRIEND
Journal of natural medicines
New glycosides of acetophenone derivatives and phenylpropanoids from Juniperus occidentalis.
PMID 22847379
Abstract
New glycosides of seven acetophenone derivatives (1-7) and two phenylpropanoids (8, 9), named juniperosides III-XI, have been isolated from the MeOH extract of the leaves and stems of Juniperus occidentalis Hook. (Cupressaceae), together with eleven other known compounds. The structures of these compounds have been successfully elucidated using a variety of spectroscopic techniques.