EMAIL THIS PAGE TO A FRIEND
Chirality
High-performance liquid chromatographic enantioseparation of unusual isoxazoline-fused 2-aminocyclopentanecarboxylic acids on (+)-(18-crown-6)-2,3,11,12-tetracarboxylic acid-based chiral stationary phases.
PMID 22847684
Abstract
The enantiomers of four unusual isoxazoline-fused 2-aminocyclopentanecarboxylic acids were directly separated on chiral stationary phases containing (+)-(18-crown-6)-2,3,11,12-tetracarboxylic acid as chiral selector. The nature of the alcoholic modifier (MeOH, EtOH, IPA) exerted a great effect on the retention, whereas the selectivity and resolution did not change substantially. Two types of dependence of retention on alcohol content were detected: k(1) increased continuously with increasing alcohol content or a U-shaped retention curve was observed. A comparison of the chromatographic data obtained with HCOOH, AcOH, TFA, HClO(4), H(2)SO(4), or H(3)PO(4) as acidic modifier at a constant concentration demonstrated that in most cases, larger k values were obtained on the application of AcOH or HCOOH, and an increase of the acid content resulted in a decrease of retention. Some mechanistic aspects of the chiral recognition process are discussed with respect to the structures of the analytes and selector. The sequence of elution of the enantiomers was determined in all cases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442666
(+)-(18-Crown-6)-2,3,11,12-tetracarboxylic acid, 97%
C16H24O14
A48105
Cycloleucine, 97%
C6H11NO2