EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Enantioselective addition of cyclic enol silyl ethers to 2-alkenoyl-pyridine-N-oxides catalysed by Cu(II)-bis(oxazoline) complexes.
PMID 22847937
Abstract
Asymmetric reactions involving (E)-3-aryl-1-(pyridin-2-yl-N-oxide)prop-2-en-1-ones and cyclic enol silyl ethers show good yields and excellent enantioselectivities (up to 99.9 % ee) when catalysed by bis(oxazoline)-Cu(II) complexes. Different reaction pathways can be followed by different enol silyl ethers: with 2-(trimethylsilyloxy)furan, a Mukaiyama-Michael adduct is obtained, whereas a hetero Diels-Alder cycloadduct was formed by using (1,2-dihydronaphthalen-4-yloxy)trimethylsilane. In the latter reaction, the absolute configuration of the product is consistent with a reagent approach to the less hindered Re face of the coordinated substrate in the reactive complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131652
Pyridine N-oxide, 95%
C5H5NO