PloS one
The obesity-related peptide leptin sensitizes cardiac mitochondria to calcium-induced permeability transition pore opening and apoptosis.
PMID 22848545
Abstract
The obesity-related 16 kDa peptide leptin is synthesized primarily in white adipocytes although its production has been reported in other tissues including the heart. There is emerging evidence that leptin may contribute to cardiac pathology especially that related to myocardial remodelling and heart failure. In view of the importance of mitochondria to these processes, the goal of the present study is to determine the effect of leptin on mitochondria permeability transition pore opening and the potential consequence in terms of development of apoptosis. Experiments were performed using neonatal rat ventricular myocytes exposed to 3.1 nM (50 ng/ml) leptin for 24 hours. Mitochondrial transition pore opening was analyzed as the capacity of mitochondria to retain the dye calcein-AM in presence of 200 µM CaCl2. Leptin significantly increased pore opening although the effect was markedly more pronounced in digitonin-permeabilized myocytes in the presence of calcium with both effects prevented by the transition pore inhibitor sanglifehrin A. These effects were associated with increased apoptosis as evidenced by increased TUNEL staining and caspase 3 activity, both of which were prevented by the transition pore inhibitor sanglifehrin A. Leptin enhanced Stat3 activation whereas a Stat 3 inhibitor peptide prevented leptin-induced mitochondrial transition pore opening as well as the hypertrophic and pro-apoptotic effects of the peptide. Inhibition of the RhoA/ROCK pathway prevented the hypertrophic response to leptin but had no effect on increased pore opening following leptin administration. We conclude that leptin can enhance calcium-mediated, Stat3-dependent pro-apoptotic effects as a result of increased mitochondrial transition pore opening and independently of its hypertrophic actions. Leptin may therefore contribute to mitochondrial dysfunction and the development of apoptosis in the diseased myocardium particularly under conditions of excessive intracellular calcium accumulation.