EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Ginsenoside Re ameliorates inflammation by inhibiting the binding of lipopolysaccharide to TLR4 on macrophages.
PMID 22849695
Abstract
Ginseng (the root of Panax ginseng C.A. Meyer, family Araliaceae), which contains protopanaxadiol ginsenoside Rb1 and protopanaxatriol ginsenoside Re as main constituents, is frequently used to treat cancer, inflammation, and stress. In the preliminary study, protopanaxatriol ginsenoside Re inhibited NF-κB activation in lipopolysaccharide (LPS)-stimulated murine peritoneal macrophages. Therefore, we investigated its anti-inflammatory effect in peptidoglycan (PGN)-, LPS-, or tumor necrosis factor-α (TNF-α)-stimulated peritoneal macrophages and, in addition, in LPS-induced systemic inflammation and 2,4,6-trinitrobenzene sulfonic acid (TNBS)-induced colitis in mice. Ginsenoside Re inhibited IKK-β phosphorylation and NF-κB activation, as well as the expression of proinflammatory cytokines, TNF-α and IL-1β, in LPS-stimulated peritoneal macrophages, but it did not inhibit them in TNF-α- or PG-stimulated peritoneal macrophages. Ginsenoside Re also inhibited IRAK-1 phosphorylation induced by LPS, as well as IRAK-1 and IRAK-4 degradations in LPS-stimulated peritoneal macrophages. Ginsenoside Re inhibited the binding of Alexa Fluor 488-conjugated LPS to TLR4 on peritoneal macrophages. Furthermore, ginsenoside Re inhibited the binding of LPS to TLR4 on peritoneal macrophages transiently transfected with MyD88 siRNAs. Orally administered ginsenoside Re significantly inhibited the expression of IL-1β and TNF-α on LPS-induced systemic inflammation and TNBS-induced colitis in mice. Ginsenoside Re inhibited colon shortening and myeloperoxidase activity in TNBS-treated mice. Ginsenoside Re reversed the reduced expression of tight-junction-associated proteins ZO-1, claudin-1, and occludin. Ginsenoside Re (20 mg/kg) inhibited the activation of NF-κB in TNBS-treated mice. On the basis of these findings, ginsenoside Re may ameliorate inflammation by inhibiting the binding of LPS to TLR4 on macrophages.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03000590
Ginsenoside Re, primary reference standard
C48H82O18