EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Heteropolyacid-catalyzed direct deoxygenation of propargyl and allyl alcohols.
PMID 22849794
Abstract
The combination of H(3)[PW(12)O(40)]·nH(2)O (1 mol %) and Et(3)SiH led to the direct catalytic deoxygenation of propargyl alcohols, in which proper solvent selection Cl(CH(2))(2)Cl vs CF(3)CH(2)OH was the key to obtaining better product yields. Under similar conditions, the deoxygenation of allyl alcohols proceeded to give thermodynamically stable alkenes with migration of the double bonds in good yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50803
Propargyl alcohol, 99%
C3H4O