EMAIL THIS PAGE TO A FRIEND
Agents and actions
Antiinflammatory activity of methionine, methionine sulfoxide and methionine sulfone.
PMID 2285015
Abstract
The oxidation of methionine in peptides is often associated with the loss of biological activity. Since methionine showed good antiinflammatory activity, its oxidized products methionine sulfoxide and methionine sulfone were tested. The sulfone was more active than the sulfoxide although methionine was most active indicating that the antiinflammatory activity is not correlated with the oxidation state of sulphur. Their hydroxyl radical scavenging activity was measured. Methionine was most active and sulfone was least active. Here also no correlation with antiinflammatory activity was found.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64410
DL-Methionine sulfone, ≥99.0% (NT)
C5H11NO4S
M0876
L-Methionine sulfone
C5H11NO4S