EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Asymmetric Brønsted acid-catalyzed nazarov cyclization of acyclic α-alkoxy dienones.
PMID 22851505
Abstract
A Brønsted acid-catalyzed asymmetric Nazarov cyclization of acyclic α-alkoxy dienones has been developed. The reaction offers access to chiral cyclopentenones in a highly enantioselective manner. The reaction is complementary to our previously reported Brønsted acid-catalyzed electrocyclization reactions, which provided differently substituted optically active cyclopentenones with a fused tetrahydropyrane ring in good yields and with excellent enantioselectivities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C112909
2-Cyclopenten-1-one, 98%
C5H6O